Key Documents
Z645206
IKA® A 11.6 double beater
A 11.6 double beater, 1/cs
About This Item
UNSPSC Code:
41121800
NACRES:
NB.74
feature
only applicable with grinding chamber A 11.4
packaging
case of 1
pack of 1
manufacturer/tradename
IKA 3302900
suitability
suitable for (use up to Mohs hardness 3)
General description
Requires A11.4 grinding chamber
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)