Z645028
IKA® MS 3 basic shakers
universal small shaker, AC/DC input 110 V / 230 V AC, universal plug set
Synonym(s):
lab shakers, laboratory shakers, microplate shakers, orbital shakers, vortexer
About This Item
Recommended Products
feature
CE compliant
packaging
pkg of 1 ea
manufacturer/tradename
IKA 361700X
W × H × D
148 mm × 63 mm × 205 mm
weight
2.9 kg
General description
Compact, universal small shaker suitable for shaking tasks with all small vessels and microtiter plates.
- Wide range of attachments
- Attachment detection
- Continuous or touch operation (with standard attachment)
- Stable in all speed ranges
- Sturdy zinc die cast casing
- Two operating modes:
Mode A (safe mode with attachment detection) the maximum speed of 3000 rpm is only reached with the standard attachment in touch mode. When using other attachments the speed is limited to 1,300 rpm.
Mode B (without attachment detection) a speed of 3000 rpm is possible with all attachments.
Kit contents:shaker MS 3 basic, power supply, standard attachment MS 3.1, universal attachment MS 3.3, one-hand insert MS 1 & operating instructions
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)