Key Documents
Z639923
KNF Laboport® vacuum systems, SCC series
KNF SCC 840
About This Item
UNSPSC Code:
12352200
Pricing and availability is not currently available.
AC/DC input
220 V AC
feature
CE compliant
CSA approved
<8 mbar final vacuum
manufacturer/tradename
KNF SCC 840
availability
not available in USA and Canada
parameter
34 L/min pump flow
Legal Information
Laboport is a registered trademark of KNF Neuberger GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)