Key Documents
Z639907
KNF Laboxact vacuum systems -SEM series
KNF SEM 840
About This Item
UNSPSC Code:
41123304
Pricing and availability is not currently available.
AC/DC input
220 V AC
feature
CE compliant
<8 mbar final vacuum
manufacturer/tradename
KNF SEM 840
availability
not available in USA and Canada
parameter
34 L/min pump flow
General description
The pump uses chemical resistant membranes and the systems are ideally suitable for use with rotary evaportors.
Complete vacuum system consisting of membrane pump unit, high performance condenser, manual control valve, manual vacuum release valve and digital vacuum meter mounted onto a base plate.
Choose from one of the most recent versions:
Certificates of Analysis (COA)