Key Documents
Z639672
Testo® datalogger accessories
ComSoft 3 software Basic
About This Item
UNSPSC Code:
12352200
Pricing and availability is not currently available.
manufacturer/tradename
Testo®
availability
not available in USA and Canada
General description
The software is designed for data retrieval, data storage and data evaluation from and programming of any testo data logger series174, 175 and 177. It will run under the following Windows versions: 95, 98, NT, ME and XP. The Pro version offers additional data management within a tree structure to fully reflect your installation.
Legal Information
Testo is a registered trademark of Testo GmbH & Co.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)