Key Documents
Z623962
Hirschmann® microcapillary pipette
manually operated
About This Item
UNSPSC Code:
41112203
Pricing and availability is not currently available.
manufacturer/tradename
Hirschmann 9609900
availability
available only in EU
General description
Manually operated pipetting device for disposable microcapillaries with ringmark with exchangeable adapters for different capillary sizes.
Legal Information
Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)