Key Documents
Z616826
Büchi® glass oven for micro distillation, B-585
temperature and time can be programmed at 3 levels
About This Item
UNSPSC Code:
41100000
AC/DC input
240 V AC
manufacturer/tradename
Büchi 046601
General description
The ball tube oven is optimally suited for the fractioned distillation of small amounts of solvents up to 35 mL and to a maximum temperature of 300°C. Visual monitoring of the sample chamber, graphic display with manual and programmable mode.
Adjustable temperature and time control, programmable for three steps. Variable rotation up to 50 U/min, vacuum and inert gas connection, integrated cooling.
Overheating and protection against contact for the heating tube. Use of the rotation drying flask for quick drying without lumps. Optional automatic pump, on/off switch on termination of the program.
Complete and ready to use including: 1 rotation drying flask 60 mL with vapor duct (DDR), 3 ball tubes 20 mL with vapor duct, 1 cooling device, diverse standard accessories and operating manual.
Features and Benefits
  • To distill small quantities of substances
  • For distilling solvents with low boiling points
  • For rotational drying variable rotation up to 50 rpm
  • Temperature and time can be programmed at 3 levels
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)