Key Documents
Z615706
Julabo® bath test tube racks
to fit 21 test tubes, diam. 30-31 mm, polypropylene
Synonym(s):
tube holder
About This Item
UNSPSC Code:
41122804
NACRES:
NB.77
material
polypropylene
feature
Holds 21 x 30-31 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970383
parameter
+80 °C temp. limit
diam.
30-31 mm
General description
Test tube racks, for 21 tubes, 30/31 mm, diameter, to +80°C. For use with shaking water bath models SW22, SW23. Requires basic tray 8970369.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)