Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z603864

Thermo Electron Corporation bath liquid

SIL100, temperature −5-75 °C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103706
Pricing and availability is not currently available.

manufacturer/tradename

SIL100

availability

not available in USA and Canada

parameter

−5-75 °C temperature

Looking for similar products? Visit Product Comparison Guide

General description

Synthetic thermal and silicone oil for use as bath liquid for circulator baths. 10L pack.

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

214.0 °F - closed cup

flash_point_c

101.1 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service