Key Documents
Z602760
Normag liquid-liquid extractor
capacity 2,000 mL, for solvents lighter than water
About This Item
UNSPSC Code:
12352200
NACRES:
NB.43
Pricing and availability is not currently available.
joint
female joint ST/NS top45/50
male joint ST/NS side arm24/40
manufacturer/tradename
Normag SAA 02073 00
availability
not available in USA and Canada
capacity
2,000 mL
General description
A rotating distributor in the extractor vessel is driven by a magnetic stirring plate. The extractor solvent fed from the condenser to the distributor is centrifugally forced through the small holes in the distributor ring as fine droplets into the liquid to be extracted, producing optimum exchange of matter. The liquid to be extracted also rotates in the extractor.
The process is continuous and extraction is performed much more quickly than in traditional equipment. The rotary distributor on the inlet tube has a magnetic agitating rod, pivot nipple, bearings, antislip lock and separating ring with retaining ring.
Components
  • Extraction vessel has 45/40 female and 24/40 male joints (A1)
  • Inlet tube has 60/46 male and 29/32 female joints and bearings (A2)
  • Rotary distributor with magnetic agitating rod, pivot nipple, and bearing (A3)
  • Antislip lock (A4)
  • Separating ring with retaining ring (A5)
Legal Information
Normag is a trademark of Normag Labor- und Prozesstechnik GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)