Key Documents
Z602655
Normag dosing funnel
capacity 50 mL
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
material
borosilicate glass 3.3
joint
joint ST/NS29/32
manufacturer/tradename
Normag
availability
not available in USA and Canada
capacity
50 mL
General description
Jacketed with interchangeable glass needle valve.
Legal Information
Normag is a trademark of Normag Labor- und Prozesstechnik GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)