Key Documents
Z569046
Sigma-Aldrich
Handbook of Solid Phase Microextraction
J. Pawliszyn, ed., Chemical Industry Press, 2009, 406 pp., hard cover
About This Item
ISBN-13:
978-7-122-04701-4
UNSPSC Code:
55101509
publication info
J. Pawliszyn, ed., Chemical Industry Press, 2009, 406 pp., hard cover
General description
This 400 page book contains comprehensive description of the fundamental principles of solid phase microextraction, recent applications, SPME devices and procedures published to date. SPME protocols are presented in a step-by-step fashion, providing useful tips and potential pitfalls. The important steps in SPME method development and optimization including calibration are clearly discussed to assist new users of the technology. This handbook enables researchers at all stages of their careers to effectively apply this convenient and solvent free sample preparation to solve their analytical challenges in effective way. This up-to date handbook contains 13 chapters.
Topics covered include:
Theory of SPME, SPME devices and fibre coatings, commercial devices and coatings, automated SPME systems, calibration of extraction step, SPME method development, ligand-receptor binding, in-vivo SPME, review of different application areas including: environmental, food and fragrance, forensic and drug analysis as well as SPME protocols. In addition the book contains a CD with SPME presentations and description of beginning experiments helpful in designing course and lectures on the topic.
Choose from one of the most recent versions:
Certificates of Analysis (COA)