Key Documents
Z569038
Sigma-Aldrich
Aldrich® Liebig condenser with cup
Overall H 300 mm, joint: ST/NS 29/32
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
joint
joint: ST/NS29/32
Overall H
300 mm
General description
Condensate cup catches water that forms on the outer surface of the condenser during use and eliminates puddling on reactor and equipment. Tubing connects to cup′s hose barb to drain away water automatically.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)