Key Documents
Z567914
Q-Block
for 12 mL Q-Tube
Synonym(s):
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
About This Item
UNSPSC Code:
41100000
manufacturer/tradename
Q Labtech
General description
These anodized aluminum heating blocks are designed for use with 12 mL and 35 mL Q-Tubes. The maximum working temperature is 204°C Blocks have a safety locking plate, a safety shield, thermally stable silicone tubing and an internal cooling channel for fast air cooling (nitrogen, compressed air or vacuum). There is also a thermocouple well for accurate temperature measurement.
Legal Information
Q-Block is a trademark of Q Labtech LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)