Key Documents
Z567760
High pressure adapter for Q-Tube
Synonym(s):
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
About This Item
UNSPSC Code:
41100000
manufacturer/tradename
Q Labtech
parameter
-200 psi pressure (max)
Legal Information
Q-Tube is a trademark of Q Labtech LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–4 of 4 Questions  
  1. What are the benefits of High pressure adapters for Q-Tube™?
    1 answer
    1. The Q-TubeTM reactors are affordable alternatives to a microwave synthesizer. Unlike the microwave system, which can experience hot-spots that can lead to charring of product, the Q-TubeTM system provides more uniform heat distribution.
      Helpful?
  2. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
      Helpful?
  3. What is the maximum liquid fill level for the High pressure adapter for Q-Tube™ glass tube?
    1 answer
    1. We would recommend keeping the level of the liquid just below half full. The 35mL system has a maximum fill line printed on the tube.
      Helpful?
  4. Can High pressure adapters for Q-Tube™ be heated with an oil bath?
    1 answer
    1. Yes. However, the Q-BlockTM heating block offers the advantage of being equipped with a safety shield, lattice clamps and hose barb for nitrogen stream to cool down the block.
      Helpful?