Z566918
IKA® RV 10 rotary evaporator, basic
diagonal condenser, plastic-coated glass, AC/DC input 115 V AC, pack of 1 ea
Synonym(s):
rotary evaporator condensers, rotovap condensers
About This Item
Recommended Products
material
plastic-coated glass
feature
CSA approved
packaging
pack of 1 ea
parameter
20-280 rpm speed
W × H × D
500 mm × 430 mm × 410 mm
bath capacity
3 L
weight
19.6 kg
Looking for similar products? Visit Product Comparison Guide
General description
The RV 10 evaporators are fitted with a water/oil bath that includes a powerful 1300 Watt heater capable of delivering temperatures from room temperature to 180°C. On the basic model, the temperature is controlled using a simple dial, and the basic system now comes with a drive capable of right-left interval operation as standard.
This produces significantly better results, especially in the case of drying processes. Highly solvent-resistant PTFE seals also ensure that operation of the system is exceptionally safe, even when used with aggressive organic solvents.
Basic model:
- The temperature is controlled using a simple dial
- Comes with a drive capable of right-left interval operation
- Solvent-resistant PTFE seals
- Supplied complete with a Woulff bottle to protect the vacuum pump against solvent condensation
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service