Z566691
BudgetSensors AFM probes
silicon tip, conductive Cr/Pt coating, mode contact
About This Item
Recommended Products
material
conductive Cr/Pt coating
silicon tip
manufacturer/tradename
Innovative Solutions ContE-10
General description
- Silicon and silicon nitride probes
- Coatings: magnetic, gold, aluminum reflex, conductive chromium/platinum, gold/chromium
- Modes: soft, tapping, force modulation, contact
Choose from one of the most recent versions:
Certificates of Analysis (COA)