Key Documents
Z566667
BudgetSensors AFM probes
silicon tip, magnetic coating,
mode force modulation
About This Item
UNSPSC Code:
41100000
material
magnetic coating
silicon tip
manufacturer/tradename
Innovative Solutions Multi75M-G-10
General description
Useful for surface analysis in materials science, chemistry, physics and biology, and semiconductor applications.
  • Silicon and silicon nitride probes
  • Coatings: magnetic, gold, aluminum reflex, conductive chromium/platinum, gold/chromium
  • Modes: soft, tapping, force modulation, contact
Choose from one of the most recent versions:
Certificates of Analysis (COA)