Key Documents
Z566454
Sigma-Aldrich
Aldrich® Cool/Heat coil for Sure/Pac station
1/4 in. I.D. hose connections
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
General description
Copper coil warms gas cylinder to maintain gas flow during dispensing. Connects to hot tap water or recirculator using 1/4 in. I.D. tubing. Coil fits 900 mL x 3 in. diameter Sure/Pac cylinders.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)