Key Documents
Z565601
Büchi® vacuum pump
V-710, AC/DC input 120 - 240 V AC
About This Item
UNSPSC Code:
41100000
feature
CE compliant
manufacturer/tradename
Büchi 072000
General description
This diaphragm vacuum pump provides excellent operating reliability and is suitable for use with the Büchi evaporators. All components that come in contact with solvent vapors are made of highly resistant materials, including PTFE membranes, and glass heads. The V-700 pump has a suction volume 1.8 m3/h and absolute vacuum of 10 mbar, making this pump suitable for many applications in the laboratory. The V-710 is a larger version with four diaphram heads and a three-stage vacuum creation process which delivers 3.1m3/h and an absolute vacuum of 2 mbar.
  • Integrated switch box automatically switches the pump on or off according to vacuum demand prolonging pump life
  • Includes silencer, waterproof cable and operating manual
  • Optional Woulff flask may be connected in front of pump as a vacuum reservior to protect pump from liquids accidentally becoming aspirated into vacuum line
  • Can be used the V-850 and V-855 vacuum controllers
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)