Key Documents
Z564273
Vacuum accessories for Buchi® vacuum pumps
Secondary cold-trap
About This Item
UNSPSC Code:
41105101
NACRES:
NB.43
manufacturer/tradename
Büchi 47190
General description
If dry ice is used instead of cooling water, a suitable cold trap can also be installed.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)