Key Documents
Z562653
Sigma-Aldrich
Open-edged fritted disc porosity assortment pack for VerSAfunnels
disc diam. 50 mm
About This Item
UNSPSC Code:
41121809
NACRES:
NB.24
disc diam.
50 mm
General description
Each pack consists of 1 of each porosity:
4-8μm
10-20μm
25-50μm
70-100μm
145-175μm
The 90mm disc pack does not include the 4-8μm porosity.
Legal Information
VerSAFunnel is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)