Key Documents
Z562572
Sigma-Aldrich
Aldrich® corrosive gas regulator with electroless nickel-plated body and cross purge assembly
inlet CGA 330
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
material
316L stainless steel diaphragm (PTFE-lined)
Monel® nozzle
description
Captured vent
PTCFE seat and Viton® seals
delivery pressure measuring range
0-80 psig
inlet pressure measuring range
3000 psig
inlet CGA
330
outlet NPTM
1/4 in.
General description
The combination of Monel® regulator components and proper purging with the cross-purge assembly makes these units virtually impervious to attack from strong acid forming gases that include hydrogen chloride, chlorine, hydrogen fluoride, hydrogen bromide, and silicon tetrafluoride.
Cross-purge assembly eliminates the escape of toxic and flammable gases when changing cylinders and permits purging system with dry, inert gas to prevent corrosion due to gas hydrolysis.
Specifications:
  • Single-stage
  • Electroless nickel-plated body
  • Stainless steel diaphragm
  • 316L Stainless steel cross-purge with color-coded 1/4 turn valves with 1/4 in. NPTM outlets
Request Technical Information Bulletin No. AL-202 for cross-purge techniques.
Other Notes
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Monel is a registered trademark of Huntington Alloys Corp.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)