Key Documents
Z561649
Aldrich® automatic shut-off quick-disconnect
flat O-ring
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
material
Viton®
O.D. × I.D. × thickness
11 mm × 5 mm × 2 mm
General description
Replace SafetyBarb tubing connectors with these new quick-disconnect fittings, which are made specifically for use with Aldrich glassware. To install quick-disconnects, unscrew tubing connectors on glassware and replace with coupling inserts Z55,337-9. Order one coupling insert and one coupling body for each glass connection. Chemically resistant acetal coupling insert and body are spring-loaded, locking, and have 316SS springs and EPR seals. When pulled apart, both sides seal quickly and automatically. For use with flexible 1/4 in. I.D. tubing.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)