Key Documents
Z560820
Sigma-Aldrich
Aldrich® Benchrack support stand and rod
with center and side mount holes
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
General description
Red powder-epoxy enamel coated steel base, 8 x 8 x 1/4 in. with center and side mount holes. Includes two 5/8 x 113/4 in. aluminum rod sections with threaded ends that screw together. Additional rod sections can be used to add extra height.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchrack is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)