Key Documents
Z560219
Sigma-Aldrich
Aldrich® Benchrack large lattice system kit
wall-mounted
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
H × W
32 in. × 26 in.
General description
The red powder-epoxy enamel coated, heavy-gauge frames of the Benchrack floor-mounted and wall-mounted lattice systems bolt together and are expandable using the add-on parts listed below each system. Systems have 3 vertical and 3 horizontal ½ in. diam. aluminum lattice rods that adjust to any position. Includes 6 machined aluminum rod supports and 9 rod clamps with stainless steel screws and Allen wrench.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchrack is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)