Key Documents
Z560103
NOWPak® pressure dispense connector for BIB, with 3/8 in. fluid line and 1/4 in. pneumatic pressure line
pkg of 1 ea
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
packaging
pkg of 1 ea
fitting
1/4 in. fitting (gas line)
3/8 in. fitting (dispense line)
General description
Includes: Tube fittings for chemical feed and gas pressure lines.
The removable FEP diptube is sized to fit a 20 L container. Wetted materials include PTFE, FEP, PFA and a Chemraz® seal. ATMI No. (ND-LD-18).
Legal Information
Chemraz is a registered trademark of Greene, Tweed of Delaware, Inc.
NOWPAK is a registered trademark of Advanced Technology Materials, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)