Key Documents
Z558141
Spinner turbines for Varian® spectrometers
variable temperature for non-spinning operation, diam. 5 mm
About This Item
UNSPSC Code:
41121705
NACRES:
SB.32
manufacturer/tradename
Wilmad V-PEEK-5-NS
diam.
5 mm
General description
Enables a portion of the VT air to pass straight past the sample inside the spinner turbine thereby reducing VT gradients and micro sonic flutter due to high VT flow.
Fiberglass turbines are made from high density GFK fiberglass and incorporate a single O-ring imbedded at the turbine top to hold the sample tube firmly into place. These spinners are suitable for Varian spectrometers and are a low cost alternative to full VT versions for sample changers.
Legal Information
Varian is a registered trademark of Varian Medical Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)