Key Documents
Z558125
Spinner turbines for Varian® spectrometers
variable temperature, PEEK, diam. 5 mm
Synonym(s):
Wilmad nmr tubes, nmr sample tubes, nmr tubes
About This Item
UNSPSC Code:
41121705
NACRES:
SB.32
manufacturer/tradename
Wilmad V-PEEK-5
diam.
5 mm
General description
Fiberglass turbines are made from high density GFK fiberglass and incorporate a single O-ring imbedded at the turbine top to hold the sample tube firmly into place. These spinners are suitable for Varian spectrometers and are a low cost alternative to full VT versions for sample changers.
VT operation of -150°C to +200°C.
Legal Information
Varian is a registered trademark of Varian Medical Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)