Key Documents
Z556874
Sigma-Aldrich
Aldrich® double jaw utility Benchclamp
nickel plated (finish), pkg of 1 ea
Select a Size
1 EA
$67.10
$67.10
Available to ship onApril 29, 2025Details
Select a Size
Change View
1 EA
$67.10
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
$67.10
Available to ship onApril 29, 2025Details
material
nickel plated (finish)
packaging
pkg of 1 ea
jaw capacity
0-76 mm , small two prong
0-95 mm , large two prong
General description
Nickel-plated finish is corrosion resistant. Clamp has a large jaw on one end and a small jaw on other.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchclamp is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Regarding your benchclamp Z556874 Aldrich® double jaw utility Benchclamp™ I want to clamp one end on 1.5" umbrella pole. Other end to 2" square balcony upright aluminum post. Will that work?
    1 answer
    1. The jaw capacity is 0-76 mm for the small two prong clamp and 0-95 mm for the large two prong clamp. This is approximately 3 in. for the small prong and 3.75 in. for the large prong. The clamp is expected to be suitable for this scenario.
      Helpful?