Key Documents
Z555150
Sigma-Aldrich
Open-edged fritted disc for Aldrich® Buchner VerSAfunnels
disc diam. 90 mm, porosity 70-100 μm
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
disc diam.
90 mm
thickness
7.5 mm
pore size
70-100 μm porosity
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
VerSAFunnel is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)