Key Documents
Z551813
Sigma-Aldrich
Aldrich® Thermometer Adapter
offset, male joint: ST/NS 19/22, GL 14 thread
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
male joint: ST/NS19/22
fitting
GL 14 thread
General description
Has PTFE faced silicone washer and open-top cap.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)