Key Documents
Z551716
Chemat environmental control chamber
transparent polycarbonate (UV resistant)
About This Item
UNSPSC Code:
41100000
material
transparent polycarbonate (UV resistant)
manufacturer/tradename
Chemat
chamber W × D × H
36 in. × 24 in. × 24 in., 1/4 in. NPT inlet and outlet ball valves, one each
door opening size
9 x 9 in. , with gasketed doors
port size
8 in. , round with flange for connecting glove
weight
48 lb (chamber)
General description
The Environmental Control Chamber is designed to provide a controllable environment for fabrication of thin films and coatings via spin coating. Hermetically sealed enclosures provide a controlled atmosphere for a wide range of fabrication and testing operations. Their modular design lets you select the options you need for your operation. All chambers are fabricated to meet rigorous standards for cleanliness and durability. Solvent welding ensures a strong, air-tight structure.
For use with the Chemat Spin-coater system.
Choose from one of the most recent versions:
Certificates of Analysis (COA)