MilliporeSigma
All Photos(6)
Key Documents
Z544604
Molymod® Individual organic model set
PVC
About This Item
UNSPSC Code:
41100000
description
monomer units 5
manufacturer/tradename
Molymod
General description
The mushroom-shaped hydrogen Atomlink components and the short (non-visible) bonding links provide compact, semi-space-filling models with an internuclear scale of 1.45 cm per Angstrom. Polymer models include long, flexible links for demonstration of the mechanism of polymerization.
Legal Information
Atomlink is a trademark of Spiring Enterprises Ltd.
Molymod is a registered trademark of Spiring Enterprises Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)