Key Documents
Z544442
Molymod® molecular model set
student set, p and pi orbital upgrade
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
description
12 p and 12 pi lobes, flat, purple/pink
manufacturer/tradename
Molymod MO-12
General description
Top quality Molymod molecular model sets are for use by students, teachers, and researchers. May be upgraded or customized with additional model parts, for more info, go to sigma-aldrich.com and enter Molymod in our search engine.
Scale is 2.8 cm =100 pm (open style) and 1.45 cm = 100 pm (compact, semi-space-filling). Sets include standard color-coded plastic atom centers (with bond holes set at the appropriate angles), bonding links, and a plastic storage box.
Set pictured is representative of the Student set.
See our Selection Guide for detailed descriptions of set contents.
Type "p" orbitals, pack of 12 (6 x pink & 6 x purple)
Legal Information
Molymod is a registered trademark of Spiring Enterprises Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)