Key Documents
Z543780
Sigma-Aldrich
Liquid-level indicator for VerSA-Flow dispenser
for 18 L poly-lined steel can
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
General description
316 SS tube and float with color-coded (blue-yellow-red) inner gauge wire for visual determination of liquid level inside of container.
Legal Information
VerSA-Flow is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)