Key Documents
Z543578
Sigma-Aldrich
Delivery cap for VerSA-Flow dispenser
three-way
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
description
Made from PBT and PTFE
O.D.
1/8 in.
3.2 mm
fitting
GL 45 female
General description
Caps require a GL-45 bottle adapter such as Z332305 for use with Aldrich 2 L and 4 L bottles.
Legal Information
VerSA-Flow is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)