Key Documents
Z543217
Sigma-Aldrich
GL-45 connector for EZSafe® flash-chromatography assembly
for attaching the solvent reservoir to the column
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
grade
for chromatography
suitability
suitable for chromatography
General description
For the attachment of the solvent reservoir to the column.
Legal Information
EZ Safe is a registered trademark of Gerresheimer Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)