General description
Lysozyme (129 residues), ribonuclease (124 residues), myoglobin (154 residues) and cytochrome C (103 residues) can be constructed from this set, which contains 160 amino acid residues (130 hydrogen-bonded amino acid backbones and 30 free amino acid backbones). Also includes labels for amino acid residues, frame parts to support the model, and supplementary parts for assembling additional residues. Scale: 1Å = 1cm.
Choose from one of the most recent versions:
Certificates of Analysis (COA)