Z531472
Aldrich® Graham condenser
Overall H 500 mm, joint: ST/NS 29/32
All Photos(1)
About This Item
UNSPSC Code:
41121800
joint
joint: ST/NS29/32
availability
not available in EU
Overall H
500 mm
General description
The Graham condenser has a large cooling surface area by allowing coolant to circulate around the inner coil. Condensers have a standard taper outer joint and a lower inner drip-tip joint.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)