Key Documents
Z530298
Sigma-Aldrich
Aldrich® straight vacuum-distilling adapter
joint: ST/NS 19/22
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
joint: ST/NS19/22
General description
With side hose connection and drip-tip. Improves efficiency of evaporation of solvents under reduced pressure when placed between the condenser of a rotary evaporator and a chilled receiving flask. Plug the condenser vacuum take-off exit opposite the receiving flask to eliminate path competition and direct all distillate to the chilled collection flask. SJ 35/25 adapter fits standard Büchi rotary evaporator glassware.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)