Key Documents
Z530166
Sigma-Aldrich
Manual gas control valve
stainless steel, CGA Inlet size 240, female ¼ in. outlet
About This Item
UNSPSC Code:
41112404
NACRES:
NB.51
material
stainless steel
inlet pressure measuring range
≤3,000 psig
CGA Inlet size
240
General description
Provides an economical means of controlling gas flow directly from the cylinder. Manufactured with SS or Monel® bodies to accommodate corrosive gases that cannot be run through a regulator. Valve stem packing is PTFE. Hose barb outlet.
Warning
Control valves do not regulate pressure, they only control flow.
Legal Information
Monel is a registered trademark of Huntington Alloys Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)