material
steel support
packaging
pkg of 1 ea
General description
- All steel construction floor mounted gas cylinder stand
- Holds one cylinder from 4.0 to 10.0 inch diameter
- All steel, finishing, packaging, and labor made 100% in the USA and Canada
- Constructed from 7 & 11 Gauge hot rolled steel
- Edges are protected with steel reinforced vinyl edge guarding to help maintain your cylinders and provide extra grip
- Steel parts are sealed with powder paint to assure long service life and chemical resistance
- Support straps are 1.5 inch wide by 54 inch long polypropylene with steel cinch buckle and rate a robust 1138 PSI and are the primary means of support
- Clamping Bar- 3/16 in. thick cut and formed steel bar with dual threaded rods and wing nuts
- Three floor mounting openings in base
Choose from one of the most recent versions:
Certificates of Analysis (COA)