Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z529745

Supelco

Aldrich® chromatography sprayer

size 250 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
NC.07

grade

for chromatography

technique(s)

thin layer chromatography (TLC): suitable

size

250 mL

suitability

suitable for chromatography

Looking for similar products? Visit Product Comparison Guide

General description

Provides a fine, uniform spray that is optimized for the development of TLC plates. Also suited for use in electrophoresis.
  • Adjustable spray pattern using thumb on vent hole
  • Greaseless, screw-threaded joint will not seize; a simple turn of the threaded cap pulls joint apart safely
  • Threaded SafetyBarb® connector prevents accidental breakage during tubing installation or removal
  • Uses low-pressure gas or air

Linkage

See the Threaded Ground Glass Tutorial to see how threaded joints work.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service