Key Documents
Z529044
Sigma-Aldrich
Aldrich® can/drum adapters
Bottle-top adapter, GL 32
About This Item
UNSPSC Code:
41121815
NACRES:
NB.01
General description
Chemically inert PBT adapters permit the attachment of pumps and bottle-top dispensers to Aldrich 5 gal cans or drums.
GL 32 to 2-in. NPT thread. Optifix® to Aldrich 5 gallon container.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Optifix is a registered trademark of Poulten & Graf Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)