Key Documents
Z528730
Sealing system for Büchi® R200/205 evaporators
adapter seal
About This Item
UNSPSC Code:
41104804
NACRES:
NB.43
feature
This product is the adapter seal
manufacturer/tradename
Büchi 40030
General description
Fits R200 and R205 evaporators. Picture shows vacuum seal on left, vapor duct in middle and adapter seal is on the right.
Fits all the glass assembly styles of the R200/205 model.
This product is the adapter seal
Other Notes
The sealing system consists of a sealing ring and an o-ring
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)