Key Documents
Z527696
Plastic drum plugs and caps
NPT threaded plug for metal drums, Rieke 2 inch, with rubber septum-inlet
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
packaging
pkg of 5 ea
General description
Rieke® plugs and caps are HDPE with an EPDM gasket. Tri-Sure plugs are PP with a rubber gasket. Satisfy FDA and UN packaging standards. Drum faucets will fit plugs that have a 3/4 in. NPT center hole.
Legal Information
Rieke is a registered trademark of Rieke Metals, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)