Key Documents
Z517593
Sigma-Aldrich
Aldrich® round-bottom storage flask with stopcock-equipped septum-inlet
capacity 500 mL, with 4 mm glass stopcock, joint: ST/NS 29/32
Synonym(s):
bottom flask, flask round, round flask
About This Item
UNSPSC Code:
41121804
material
borosilicate glass
round bottom flask
single-neck
joint
joint: ST/NS29/32
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
500 mL
General description
With ground-glass joint. Use septum Z565695 or Z565679 (see Septa Section).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)