Key Documents
Z517526
Sigma-Aldrich
Aldrich® vacuum manifold with joints
positions, 3, joint: ST/NS 29/32
About This Item
UNSPSC Code:
40161501
NACRES:
NB.79
joint
joint: ST/NS29/32
feature
positions 3
L
300 mm
General description
This practical design provides clearance to accommodate 250mL flasks with a snap of a Keck® clip. Manifolds are available with either <ST/>14/20, <ST/>24/40, or <ST/>29/32 joints. Glass stopcocks have a 4mm bore.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Keck is a registered trademark of Adcuram Lab Technologies GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)