Key Documents
Z517062
Sigma-Aldrich
Aldrich® Snyder distilling column
Joints: ST/NS 14/20 (2), four sections
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
joint
Joints: ST/NS14/20 (2)
I.D. × O.D.
9 mm × 13 mm
effective L
225 mm
General description
Floating ball type with joints. Highly efficient column for atmospheric distillations. Effectively separates close boiling materials. One ball per section.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)